Check Order Status
Gold Standard Metal Solution 1000ppm, for AAS, Solution HCl 1M, Fisher Chemical
Element Search
Structure Search
Gold Standard Metal Solution 1000ppm, for AAS, Solution HCl 1M, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCJ802808
also known as FGCJ/8028/08
500 mL
Catalog number
FGCJ802808
also known as FGCJ/8028/08
Price (MYR)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Packaging
HDPE plastic bottle
Physical Form
Liquid
MDL Number
3436
Color
Colorless
Quantity
500 mL
View more