Check Order Status
Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Actual product may vary
Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical, Quantity: 1AMP, Packaging: Ampoule, Boiling Point: 100 deg.C, Colorless, MDL Number: 3499, Physical Form: Liquid
Have Questions?
Catalog Number
Quantity
FGCJ7950C05
also known as FGCJ/7950C/05
1 Ampule
Catalog number
FGCJ7950C05
also known as FGCJ/7950C/05
Price (MYR)
353.00
EA
Add to cart
Quantity:
1 Ampule
Price (MYR)
353.00
EA
Add to cart
Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical, Quantity: 1AMP, Packaging: Ampoule, Boiling Point: 100 deg.C, Colorless, MDL Number: 3499, Physical Form: Liquid
For volumetric analysis
Specifications
Color
Colorless
CAS
7772-98-7
Boiling Point
100°C
Molecular Formula
Na2O3S2
MDL Number
MFCD00003499
InChI Key
AKHNMLFCWUSKQB-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]S([S-])(=O)=O
IUPAC Name
disodium pentahydrate sulfanidesulfonate
Molecular Weight (g/mol)
158.10
Packaging
Ampoule
Quantity
1 Ampule
Physical Form
Liquid