Check Order Status
Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis
Element Search
Structure Search
Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis
Have Questions?
Change view
Quantity:
2.5 L
1 L
Packaging:
HDPE plastic bottle
Catalog number
FGCJ780017
also known as FGCJ/7800/17
Price (MYR)
323.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
323.00
EA
Add to cart
Specifications
pH
12
Boiling Point
120°C
CAS
1310-73-2
Melting Point
8°C
Molecular Formula
HNaO
View more
Sodium Hydroxide Solution 10M (10N) ≈40 (w/v), Standard Solution Ready To Use, for Volumetric Analysis