Check Order Status
Sodium Hydroxide Solution Methanolic 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Element Search
Structure Search
Sodium Hydroxide Solution Methanolic 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCJ762517
also known as FGCJ/7625/17
2.5 L
Catalog number
FGCJ762517
also known as FGCJ/7625/17
Price (MYR)
-
Quantity:
2.5 L
Request bulk or custom format
Specifications
Color
Colorless
CAS
1310-73-2
Molecular Formula
HNaO
MDL Number
3548
Synonym
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
View more