Potassium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical
Potassium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical
Potassium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical
Potassium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical
Potassium Hydroxide Solution 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical
Have Questions?
Catalog NumberQuantity
FGCJ6630PB15
also known as FGCJ/6630/PB15
1 L
Catalog number FGCJ6630PB15
also known as FGCJ/6630/PB15
Price (MYR)
500.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
500.00
EA
Add to cart
Specifications
pH13
CAS1310-58-3
Molecular FormulaHKO
MDL NumberMFCD00003553
Synonympotassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
View more
Ready to use; For volumetric analysis