Check Order Status
Nitric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Element Search
Structure Search
Nitric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FGCJ5600PB15
also known as FGCJ/5600/PB15
1 L
HDPE plastic bottle
Catalog number
FGCJ5600PB15
also known as FGCJ/5600/PB15
Price (MYR)
578.00
EA
Add to cart
Quantity:
1 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
578.00
EA
Add to cart
Specifications
Color
Colorless
CAS
7697-37-2
Packaging
HDPE plastic bottle
Quantity
1 L
Molecular Formula
HNO3
View more