Check Order Status
Nitric Acid Solution 2M (2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical
Element Search
Structure Search
Nitric Acid Solution 2M (2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCJ5545PB17
also known as FGCJ/5545/PB17
2.5 L
HDPE plastic bottle
Catalog number
FGCJ5545PB17
also known as FGCJ/5545/PB17
Price (MYR)
622.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Price (MYR)
622.00
EA
Add to cart
Specifications
pH
1
CAS
7697-37-2
Boiling Point
100°C
Packaging
HDPE plastic bottle
Quantity
2.5 L
View more