Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
ClH, CAS Number-7647-01-0, spirits of salt, hydrochloric acid, chlorohydric acid, chloorwaterstof, hydrogen chloride, chlorwasserstoff, hydrogen chloride hcl, muriatic acid, anhydrous hydrochloric acid, acide chlorhydrique, 6AMP, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
6 Ampules
1 Ampule
Catalog number FGCJ4350C90
also known as FGCJ/4350C/90
Price (MYR)
689.00
EA
Add to cart
Quantity:
6 Ampules
Request bulk or custom format
Price (MYR)
689.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydrochloric Acid
ColorColorless
CAS7647-01-0
Boiling Point100°C
PackagingAmpoule
View more
ClH, CAS Number-7647-01-0, spirits of salt, hydrochloric acid, chlorohydric acid, chloorwaterstof, hydrogen chloride, chlorwasserstoff, hydrogen chloride hcl, muriatic acid, anhydrous hydrochloric acid, acide chlorhydrique, 6AMP, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol