Check Order Status
Hydrochloric Acid Solution 0.2M (0.2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Element Search
Structure Search
Hydrochloric Acid Solution 0.2M (0.2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCJ434015
also known as FGCJ/4340/15
1 L
Catalog number
FGCJ434015
also known as FGCJ/4340/15
Price (MYR)
324.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
324.00
EA
Add to cart
Specifications
pH
2
Color
Colorless
CAS
7647-01-0
Boiling Point
100°C
Packaging
HDPE plastic bottle
View more
Hydrochloric Acid Solution 0.2M (0.2N), NIST Standard Solution Ready To Use, for Volumetric Analysis