Check Order Status
Formic Acid Solution 0.25M, Certified, for HPLC, Fisher Chemical
Element Search
Structure Search
Formic Acid Solution 0.25M, Certified, for HPLC, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCJ4200PB08
also known as FGCJ/4200/PB08
500 mL
Plastic Coated Glass bottle
Catalog number
FGCJ4200PB08
also known as FGCJ/4200/PB08
Price (MYR)
-
Quantity:
500 mL
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Specifications
CAS
64-18-6
Color
Colorless
Packaging
Plastic Coated Glass bottle
Molecular Formula
CH2O2
MDL Number
3297
View more