Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.1M (0.2N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP , Fisher Chemical
Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.1M (0.2N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP , Fisher Chemical
Actual product may vary
Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.1M (0.2N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP , Fisher Chemical
Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.1M (0.2N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP , Fisher Chemical
Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.1M (0.2N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP , Fisher Chemical
Have Questions?
Catalog NumberQuantityPackaging
FGCJ370015
also known as FGCJ/3700/15
1 LHDPE plastic bottle
Catalog number FGCJ370015
also known as FGCJ/3700/15
Price (MYR)
282.00
EA
Add to cart
Quantity:
1 L
Packaging:
HDPE plastic bottle
Price (MYR)
282.00
EA
Add to cart
1L, HDPE plastic bottle
Ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP
Specifications
CAS139-33-3
ColorColorless
PackagingHDPE plastic bottle
Molecular FormulaC10H14N2Na2O8
MDL NumberMFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
Quantity1 L
Synonymethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
InChI KeyZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
IUPAC Namedisodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
Molecular Weight (g/mol)336.21
PubChem CID57339238
ChEBICHEBI:64734
Physical FormLiquid
Unit SizeEA