Check Order Status
Potassium Chloride Solution, 3M, Electrode Filling Solution, Saturated with Silver Chloride, Fisher Chemical™
Element Search
Structure Search
Potassium Chloride Solution, 3M, Electrode Filling Solution, Saturated with Silver Chloride, Fisher Chemical™
Saturated potassium chloride electrode solution suitable for use as an electrode filling solution.
Have Questions?
Catalog Number
Quantity
FGCJ289407
also known as FGCJ/2894/07
250 mL
Catalog number
FGCJ289407
also known as FGCJ/2894/07
Price (MYR)
-
Quantity:
250 mL
Request bulk or custom format
Specifications
Physical Form
Liquid
Quantity
250 mL
Packaging
HDPE Plastic Bottle