Check Order Status
Ammonium Thiocyanate Solution 0.1M (0.1N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Element Search
Structure Search
Ammonium Thiocyanate Solution 0.1M (0.1N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCJ1500PB17
also known as FGCJ/1500/PB17
2.5 L
Catalog number
FGCJ1500PB17
also known as FGCJ/1500/PB17
Price (MYR)
638.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
638.00
EA
Add to cart
Specifications
Color
Colorless
pH
4.5 to 6.0
Physical Form
Liquid
Quantity
2.5 L
MDL Number
11428
View more