Check Order Status
Ammonia Solution 0.25M, Certified, for HPLC, Fisher Chemical
Element Search
Structure Search
Ammonia Solution 0.25M, Certified, for HPLC, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCJ0100PB08
also known as FGCJ/0100/PB08
500 mL
Catalog number
FGCJ0100PB08
also known as FGCJ/0100/PB08
Price (MYR)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
pH
12
Color
Colorless
Boiling Point
100°C
CAS
1336-21-6
Molecular Formula
H5NO
View more