Check Order Status
N-2-Hydroxyethylpiperazine-N'-2-Ethanesulfonic Acid, for Biochemistry, Fisher Chemical
Element Search
Structure Search
N-2-Hydroxyethylpiperazine-N'-2-Ethanesulfonic Acid, for Biochemistry, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCH223048
also known as FGCH/2230/48
100 g
Plastic powder jar
Catalog number
FGCH223048
also known as FGCH/2230/48
Price (MYR)
-
Quantity:
100 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
CAS
7365-45-9
Melting Point
234°C
Color
White
pH
5.0 to 6.5
Packaging
Plastic powder jar
View more