Check Order Status
Hydriodic Acid, 64-68%, Certified AR for Analysis, Fisher Chemical
Element Search
Structure Search
Hydriodic Acid, 64-68%, Certified AR for Analysis, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCH085007
also known as FGCH/0850/07
250 mL
Amber glass bottle
Catalog number
FGCH085007
also known as FGCH/0850/07
Price (MYR)
10,584.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Amber glass bottle
Request bulk or custom format
Price (MYR)
10,584.00
EA
Add to cart
Specifications
pH
1
Color
Brown-Red
CAS
10034-85-2
Melting Point
-51°C
Boiling Point
127°C
View more