Hexanes, for HPLC, 95% n-Hexane approx., Fisher Chemical
Hexanes, for HPLC, 95% n-Hexane approx., Fisher Chemical
Hexanes, for HPLC, 95% n-Hexane approx., Fisher Chemical
Hexanes, for HPLC, 95% n-Hexane approx., Fisher Chemical
Hexanes, for HPLC, 95% n-Hexane approx., Fisher Chemical
High purity and versatile solvent suitable for HPLC, spectrophotometry, and GC/MS analysis
Have Questions?
Catalog number FGCH040617
also known as FGCH/0406/17
Price (MYR)
532.00
BOT
Add to cart
Request bulk or custom format
Price (MYR)
532.00
BOT
Add to cart
Specifications
Melting Point-95°C
CAS110-54-3
ColorColorless
Boiling Point69°C
Quantity2.5 L
View more
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested for GC/MS suitability
  • Also meets ACS specifications
  • Intended for research use only