Check Order Status
Isohexane, contains <5% N-Hexane, for HPLC
Element Search
Structure Search
Isohexane, contains <5% N-Hexane, for HPLC
Have Questions?
Catalog Number
Quantity
Packaging
FGCH0405PB17
also known as FGCH/0405/PB17
2.5 L
Plastic-coated Glass Bottle
Catalog number
FGCH0405PB17
also known as FGCH/0405/PB17
Price (MYR)
475.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Request bulk or custom format
Price (MYR)
475.00
EA
Add to cart
Specifications
Melting Point
-154°C
CAS
73513-42-5
Color
Colorless
Boiling Point
60°C
Quantity
2.5 L
View more