Check Order Status
1,1,1,3,3,3-Hexafluoropropan-2-ol, 99+%, for Gas Chromatography, Fisher Chemical
Element Search
Structure Search
1,1,1,3,3,3-Hexafluoropropan-2-ol, 99+%, for Gas Chromatography, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCH028504
also known as FGCH/0285/04
25 mL
Amber glass bottle
Catalog number
FGCH028504
also known as FGCH/0285/04
Price (MYR)
-
Quantity:
25 mL
Packaging:
Amber glass bottle
Request bulk or custom format
Specifications
CAS
920-66-1
Melting Point
-4°C
Color
Colorless
pH
3 to 4
Boiling Point
59°C
View more