Check Order Status
N-Heptane, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
N-Heptane, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FGCH015515
also known as FGCH/0155/15
1 L
Amber Glass Bottle
Catalog number
FGCH015515
also known as FGCH/0155/15
Price (MYR)
371.00
EA
Add to cart
Quantity:
1 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
371.00
EA
Add to cart
Specifications
Melting Point
-91°C
CAS
142-82-5
Color
Colorless
Boiling Point
98°C
Quantity
1 L
View more