Check Order Status
Formamide, for Electrophoresis, Fisher Chemical™
Element Search
Structure Search
Formamide, for Electrophoresis, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCFP565PB08
also known as FGCF/P565/PB08
500 mL
Catalog number
FGCFP565PB08
also known as FGCF/P565/PB08
Price (MYR)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
pH
4 to 5
Melting Point
2°C
CAS
75-12-7
Boiling Point
210°C
Quantity
500 mL
View more