Check Order Status
Formic Acid, 98-100%, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Formic Acid, 98-100%, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCF1850PB08
also known as FGCF/1850/PB08
500 mL
Catalog number
FGCF1850PB08
also known as FGCF/1850/PB08
Price (MYR)
380.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
380.00
EA
Add to cart
Specifications
pH
2.1
CAS
64-18-6
Melting Point
8°C
Boiling Point
101°C
Color
Colorless
View more