Check Order Status
Ethidium™ Bromide, for Electrophoresis, Fisher Chemical
Element Search
Structure Search
Ethidium™ Bromide, for Electrophoresis, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCEP70044
also known as FGCE/P700/44
5 g
Glass Jar
Catalog number
FGCEP70044
also known as FGCE/P700/44
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass Jar
Request bulk or custom format
Specifications
CAS
1239-45-8
Melting Point
260°C
Color
Purple
pH
4.0 to 7.0
Packaging
Glass Jar
View more