Check Order Status
2-Ethoxyethanol, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
2-Ethoxyethanol, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Packaging:
Amber glass bottle
Catalog number
FGCE075017
also known as FGCE/0750/17
Price (MYR)
572.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber glass bottle
Request bulk or custom format
Price (MYR)
572.00
EA
Add to cart
Specifications
CAS
110-80-5
Melting Point
-100°C
Color
Colorless
pH
7
Boiling Point
135°C
View more