Check Order Status
Dithio-Oxamide, Pure, Fisher Chemical
Element Search
Structure Search
Dithio-Oxamide, Pure, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCD515046
also known as FGCD/5150/46
25 g
Plastic powder jar
Catalog number
FGCD515046
also known as FGCD/5150/46
Price (MYR)
-
Quantity:
25 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
CAS
79-40-3
Melting Point
200°C
Color
Red
Packaging
Plastic powder jar
Molecular Formula
C2H4N2S2
View more