Check Order Status
1,4-Dioxane, Extra Pure, SLR, Stabilised with BHT, Fisher Chemical
Element Search
Structure Search
1,4-Dioxane, Extra Pure, SLR, Stabilised with BHT, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCD4500PB17
also known as FGCD/4500/PB17
2.5 L
Plastic-coated Glass Bottle
Catalog number
FGCD4500PB17
also known as FGCD/4500/PB17
Price (MYR)
538.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Request bulk or custom format
Price (MYR)
538.00
EA
Add to cart
Specifications
pH
6 to 8
Melting Point
12°C
CAS
123-91-1
Color
Colorless
Boiling Point
101°C
View more