Check Order Status
DimethylFormamide, Gel Permeation Chromatography (GPC), Fisher Chemical
Element Search
Structure Search
DimethylFormamide, Gel Permeation Chromatography (GPC), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCD3847PB17
also known as FGCD/3847/PB17
2.5 L
Plastic Coated Glass bottle
Catalog number
FGCD3847PB17
also known as FGCD/3847/PB17
Price (MYR)
417.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Price (MYR)
417.00
EA
Add to cart
Specifications
CAS
68-12-2
Melting Point
-61°C
Color
Colorless
Boiling Point
153°C
Packaging
Plastic Coated Glass bottle
View more