Check Order Status
Diethyl Ether, Extra Dry, for Synthesis, Stabilised with Ethanol, Fisher Chemical™
Element Search
Structure Search
Diethyl Ether, Extra Dry, for Synthesis, Stabilised with Ethanol, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCD250215
also known as FGCD/2502/15
1 L
Catalog number
FGCD250215
also known as FGCD/2502/15
Price (MYR)
-
Quantity:
1 L
Request bulk or custom format
Specifications
Melting Point
-116°C
CAS
60-29-7
Color
Colorless
Boiling Point
34.6°C
Quantity
1 L
View more