Check Order Status
Diethyl Ether, Extra Pure, SLR, Stabilised with BHT, Fisher Chemical
Element Search
Structure Search
Diethyl Ether, Extra Pure, SLR, Stabilised with BHT, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCD240017
also known as FGCD/2400/17
2.5 L
Amber Glass Bottle
Catalog number
FGCD240017
also known as FGCD/2400/17
Price (MYR)
413.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
413.00
EA
Add to cart
Specifications
Melting Point
-116°C
CAS
60-29-7
Color
Colorless
Boiling Point
34.6°C
Quantity
2.5 L
View more