Check Order Status
Ethylenediaminetetraacetic Acid, Tetrasodium Salt Tetrahydrate, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
Ethylenediaminetetraacetic Acid, Tetrasodium Salt Tetrahydrate, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog number
FGCD075050
also known as FGCD/0750/50
Price (MYR)
-
Request bulk or custom format
Specifications
Melting Point
300°C
CAS
13235-36-4
Color
White
pH
11 to 12
Physical Form
Solid
View more