Check Order Status
Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FGCD065048
also known as FGCD/0650/48
100 g
Plastic powder jar
Catalog number
FGCD065048
also known as FGCD/0650/48
Price (MYR)
273.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic powder jar
Request bulk or custom format
Price (MYR)
273.00
EA
Add to cart
Specifications
Melting Point
252°C
CAS
6381-92-6
Color
White
pH
4 to 6
Physical Form
Solid
View more