Check Order Status
Ethylenediaminetetraacetic Acid, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Ethylenediaminetetraacetic Acid, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCD045053
also known as FGCD/0450/53
500 g
Catalog number
FGCD045053
also known as FGCD/0450/53
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Melting Point
220°C
CAS
60-00-4
Color
White
pH
2.5
Physical Form
Powder/Solid
View more