Check Order Status
Dextran, Pure, 150, M.W. 100,000-200,000, Fisher Chemical
Element Search
Structure Search
Dextran, Pure, 150, M.W. 100,000-200,000, Fisher Chemical
Have Questions?
Catalog number
FGCD012248
also known as FGCD/0122/48
Price (MYR)
-
Request bulk or custom format
Chemical Identifiers
CAS
9004-54-0
Specifications
Color
White
CAS
9004-54-0
Physical Form
Solid
MDL Number
130935
Quantity
100 g
View more