Check Order Status
Cyclohexane, for HPLC, Fisher Chemical™
Element Search
Structure Search
Cyclohexane, for HPLC, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
1 L
Packaging:
View all
Plastic-coated Glass Bottle
Catalog number
FGCC8936PB17
also known as FGCC/8936/PB17
Price (MYR)
571.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic-coated Glass Bottle
Request bulk or custom format
Price (MYR)
571.00
EA
Add to cart
Specifications
Melting Point
6.5°C
CAS
110-82-7
Color
Colorless
Boiling Point
81°C
Quantity
2.5 L
View more