Check Order Status
Copper(II) oxide, pure, wire form, Fisher Chemical™
Element Search
Structure Search
Copper(II) oxide, pure, wire form, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCC832053
also known as FGCC/8320/53
500 g
Catalog number
FGCC832053
also known as FGCC/8320/53
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Color
Black
pH
7
Melting Point
1326°C
Quantity
500 g
Packaging
Plastic powder jar
View more