Check Order Status
O-Cresol, 98-100%, Pure, Fisher Chemical
Element Search
Structure Search
O-Cresol, 98-100%, Pure, Fisher Chemical
Have Questions?
Catalog number
FGCC7320PB08
also known as FGCC/7320/PB08
Price (MYR)
876.00
Price (MYR)
876.00
EA
Add to cart
Specifications
CAS
95-48-7
Melting Point
30°C
Color
Amber
pH
4.8
Boiling Point
191°C
View more