Check Order Status
Cinnamaldehyde, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
Cinnamaldehyde, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog number
FGCC609808
also known as FGCC/6098/08
Price (MYR)
-
Request bulk or custom format
Specifications
CAS
104-55-2
Color
Yellow
Packaging
Amber glass bottle
MDL Number
7000
Physical Form
Viscous Liquid
View more