Check Order Status
2-Chlorophenol, Extra Pure, SLR, Fisher Chemical
Element Search
Structure Search
2-Chlorophenol, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog number
FGCC5040PB08
also known as FGCC/5040/PB08
Price (MYR)
-
Request bulk or custom format
Specifications
CAS
95-57-8
Melting Point
8°C
Color
Yellow
Boiling Point
173°C
Packaging
Plastic Coated Glass bottle
View more