Check Order Status
Chloroform, for HPLC, Stabilised with Amylene, Fisher Chemical
Element Search
Structure Search
Chloroform, for HPLC, Stabilised with Amylene, Fisher Chemical
Have Questions?
Catalog number
FGCC4966PB17
also known as FGCC/4966/PB17
Price (MYR)
356.00
Price (MYR)
356.00
EA
Add to cart
Specifications
Melting Point
-63°C
Color
Colorless
Boiling Point
61°C
Quantity
2.5 L
Packaging
Plastic-coated Glass Bottle
View more