Check Order Status
2-Chloroethanol, 99+%, Pure, Fisher Chemical
Element Search
Structure Search
2-Chloroethanol, 99+%, Pure, Fisher Chemical
Have Questions?
Catalog number
FGCC4880PB08
also known as FGCC/4880/PB08
Price (MYR)
-
Request bulk or custom format
Specifications
CAS
107-07-3
Melting Point
-63°C
Boiling Point
129°C
Packaging
Plastic Coated Glass bottle
Molecular Formula
C
2
H
5
ClO
View more