Check Order Status
1-Chlorobutane, for HPLC, Fisher Chemical
Element Search
Structure Search
1-Chlorobutane, for HPLC, Fisher Chemical
Have Questions?
Catalog number
FGCC475615
also known as FGCC/4756/15
Price (MYR)
486.00
Price (MYR)
486.00
EA
Add to cart
Specifications
CAS
109-69-3
Melting Point
-123°C
Color
Colorless
Boiling Point
77°C
Packaging
Amber glass bottle
View more