Check Order Status
Chlorobenzene, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Chlorobenzene, Extra Pure, SLR, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FGCC468015
also known as FGCC/4680/15
1 L
Amber Glass Bottle
Catalog number
FGCC468015
also known as FGCC/4680/15
Price (MYR)
360.00
EA
Add to cart
Quantity:
1 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
360.00
EA
Add to cart
Specifications
Melting Point
-45°C
CAS
108-90-7
Boiling Point
131°C
Quantity
1 L
Packaging
Amber Glass Bottle
View more