Check Order Status
Chloroauric Acid, Extra Pure, about 51% Au, SLR, Fisher Chemical
Element Search
Structure Search
Chloroauric Acid, Extra Pure, about 51% Au, SLR, Fisher Chemical
Have Questions?
Catalog number
FGCC464043
also known as FGCC/4640/43
Price (MYR)
-
Request bulk or custom format
Specifications
Melting Point
30°C
CAS
16903-35-8
Color
Orange
Physical Form
Solid
Quantity
1 g
View more