Check Order Status
Charcoal, Technical, ActIVated, Acid Washed, Fisher Chemical
Element Search
Structure Search
Charcoal, Technical, ActIVated, Acid Washed, Fisher Chemical
Have Questions?
Catalog number
FGCC408050
also known as FGCC/4080/50
Price (MYR)
-
Request bulk or custom format
Specifications
CAS
7440-44-0
Melting Point
3652°C
Color
Black
Packaging
Plastic powder jar
Molecular Formula
C
View more
250g, Plastic powder jar