Check Order Status
Charcoal, Technical, SLR, Lump, Wood, Fisher Chemical
Element Search
Structure Search
Charcoal, Technical, SLR, Lump, Wood, Fisher Chemical
Have Questions?
Catalog number
FGCC402060
also known as FGCC/4020/60
Price (MYR)
-
Request bulk or custom format
Specifications
CAS
7440-44-0
Color
Black
Packaging
Plastic powder jar
Molecular Formula
C
MDL Number
133992
View more