Check Order Status
Cetyl Trimethylammonium Bromide, Pure, Fisher Chemical™
Element Search
Structure Search
Cetyl Trimethylammonium Bromide, Pure, Fisher Chemical™
C19H42BrN, CAS Number-57-09-0, 500g, White, 364.45g/mol, 230 deg.C, 11772, Plastic powder jar, 5 to 7, Powder Solid
Have Questions?
Catalog number
FGCC396053
also known as FGCC/3960/53
Price (MYR)
832.00
BOT
Add to cart
Request bulk or custom format
Price (MYR)
832.00
BOT
Add to cart
Specifications
Melting Point
230°C
CAS
57-09-0
Color
White
pH
5 to 7
Physical Form
Powder/Solid
View more
C19H42BrN, CAS Number-57-09-0, 500g, White, 364.45g/mol, 230 deg.C, 11772, Plastic powder jar, 5 to 7, Powder Solid