Check Order Status
Calcium Hydride, Technical, Fisher Chemical
Element Search
Structure Search
Calcium Hydride, Technical, Fisher Chemical
Have Questions?
Catalog number
FGCC162048
also known as FGCC/1620/48
Price (MYR)
-
Request bulk or custom format
Specifications
Melting Point
675°C
CAS
7789-78-8
Color
Gray
Physical Form
Solid
Quantity
100 g
View more