OPTIZYME™ DNase I (RNase-Free), Fisher BioReagents™
OPTIZYME™ DNase I (RNase-Free), Fisher BioReagents™
Actual product may vary
OPTIZYME™ DNase I (RNase-Free), Fisher BioReagents™
Have Questions?
Catalog NumberQuantity
FGCBP8107-11000 U
Catalog number FGCBP8107-1
Price (MYR)
-
Quantity:
1000 U
OPTIZYME™ DNase I is a recombinant endonuclease that digests DNA and is RNase-Free.
Applications and Features:
  • OPTIZYME DNase I, RNase-Free is used for the preparation of DNA-free RNA
  • Removal of template DNA following in vitro transcription
  • DNA labeling by nick translation in conjunction with DNA Polymerase I
  • Preparation of DNA-free RNA prior to RT-PCR
  • Studies of DNA-protein interactions by DNase I, RNase-free footprinting
  • Enzyme activity is strictly dependent on Ca2+ and is activated by Mg2+or Mn2+ions
  • Purified from non-animal host with a lower level of intrinsic RNases
Supplied with:
  • 10X OPTIZYME DNase I Buffer: 100mM Tris-HCl (pH 7.5 at 25°C), 25mM MgCl2, 1mM CaCl2
  • 50mM OPTIZYME EDTA (for inactivation by heat at 65°C for 10min)
Source: E. Coli cells with a cloned gene encoding bovine DNase I
Quality Control:
  • Ribonuclease-free
  • Functionally tested for the digestion of template DNA after in vitro transcription
Specifications
Concentration1 U/μL
ColorColorless
pH7.5
For Use With (Application)Preparation of DNA-free RNA, removal of template DNA following in-vitro transcription, DNA labeling by nick translation in conjunction with DNA Polymerase I, Preparation of DNA-free RNA prior to RT-PCR, Studies of DNA-Protein interactions by DNase I, RNase-free footprinting
Storage BufferOPTIZYME™ DNase I Buffer 10X : Tris HCl 100mM (7.5 pH at 25°C), MgCl2 25mM, CaCl2 1mM, OPTIZYME™ EDTA 50mM (for inactivation by heat at 65°C for 10min.)
SourceE. coli cells with a cloned gene encoding bovine DNase I
Quantity1000 U
Product TypeDNase I (RNase-free)
FormLiquid
Unit SizeEA
Contents & Storage
-20°C