Check Order Status
Boric Acid, for Electrophoresis, Fisher Chemical™
Element Search
Structure Search
Boric Acid, for Electrophoresis, Fisher Chemical™
CAS: 10043-35-3 | H
3
BO
3
Have Questions?
Catalog Number
Quantity
Packaging
FGCBP56060
also known as FGCB/P560/60
1 kg
Plastic powder jar
Catalog number
FGCBP56060
also known as FGCB/P560/60
Price (MYR)
-
Quantity:
1 kg
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
pH
3.8 to 4.8
Color
White
CAS
10043-35-3
Melting Point
169°C
Packaging
Plastic powder jar
View more
Boric Acid, for Electrophoresis