Check Order Status
Molecular Sieves, Grade 514 (Type 4A; 8-12 mesh), Fisher BioReagents
Element Search
Structure Search
Molecular Sieves, Grade 514 (Type 4A; 8-12 mesh), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
FGCBP2631-500
500 g
Catalog number
FGCBP2631-500
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS
1318-02-1, 14808-60-7
Specifications
Chemical Name or Material
Molecular Sieves, Grade 514
Color
Beige
Density
2.1 g/cm
3
Identification
Pass Test
Mesh Size
8 to 12 Mesh
View more
Used for dehydration of liquids.